Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

1.04317

Millipore

Tryptic Soy Agar - Dehydrated Culture Media

NutriSelect® Prime, EP, USP, JP, ISO, FDA BAM, powder, Suitable for use in Halal and Kosher certified processes

Synonym(s):

TSA, soy agar, CASO agar, Casein-peptone Soymeal-peptone Agar, TSA medium, soybean casein digest agar, tryptic soy (caso) agar, tryptic soy agar dehydrated, Tryptone Soy Agar, Casein Soya Agar, Soybean Casein digest Agar, CASO Agar, Soybean Casein digest Agar, TSA, Tryptone Soya Agar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$348.00

$348.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
500 G
$348.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

$348.00


Please contact Customer Service for Availability

Request more information

Agency

EP
ISO
JP
USP

Quality Level

reg. compliance

FDA BAM

form

powder

manufacturer/tradename

NutriSelect® Prime

technique(s)

microbiological culture: suitable

pH

7.3 ± 0.2 (25.0 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

NutriSelect® prime Tryptic Soy Agar halal is designed as a combination of two peptones, enzymatic digest of casein and of soy-bean, which provides a high nutrition by supplying organic nitrogen, amino acids, and longer-chained peptides. The osmotic balance is maintained by sodium chloride and agar-agar is the solidifying agent.

Application

NutriSelect® prime Tryptic Soy Agar halal is used to cultivate and isolate fastidious or non-fastidious microorganisms from test samples. It is suitable for the growth of both aerobic and anaerobic microflora.

Legal Information

NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Halal-certified culture media prevent non-halal ingredient contamination, enabling microbiological QC testing without losing certification.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service