Skip to Content
MilliporeSigma
All Photos(3)

Documents

1.16974

Supelco

Peroxide Test Strips, reflectometric

0.2-20.0 mg/L (H₂O₂), REFLECTOQUANT®

Synonym(s):

Peroxide chemical test strips, Peroxide test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NB.21

product name

Peroxide Test, reflectometric, 0.2-20.0 mg/L (H2O2), for use with REFLECTOQUANT®

description

quantitative

Quality Level

product line

Reflectoquant®

specific analyte(s)

hydrogen peroxide

reaction suitability

reaction type: enzymatic reaction
reagent type: oxidant

measuring range

0.2-20.0 mg/L (H2O2)

compatibility

RQflex® 10 Plus
RQflex® 10
RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 0.2 - 20.0 mg/l H₂O₂ Reflectoquant®
Peroxide is an all purpose disinfectant which, due to its excellent antiviral, bactericidal and fungicidal action, is widely used in the food industry. Here, the Reflectoquant test strips are suited for the determination of inorganic peroxides in aqueous solutions. Furthermore the test can be used in disinfecting and rinsing solutions (e.g. in food technology, laundries) and in swimming-pool water. Peroxidase transfers peroxide oxygen to an organic redox indicator. This produces a blue oxidation product that is determined reflectometrically on the RQflex. For pre-testing a sample in an economic and quick way to estimate the peroxide concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for peroxide.

Application


  • Application of copper (I) selective ligands for PET imaging of reactive oxygen species: This research uses peroxide detection in biomedical imaging to trace reactive oxygen species in vivo, aiding in the diagnosis and study of diseases associated with oxidative stress (Tada et al., 2024).


  • Optimization of Pressurized Liquid Extraction (PLE) Parameters for Extraction of Bioactive Compounds: This study employs peroxide tests in the extraction of bioactive compounds from plant materials, enhancing the efficiency of natural product extraction processes (Chatzimitakos et al., 2024).


  • Highly Sensitive, Easy-to-Use, One-Step Detection of Peroxide-, Nitrate- and Chlorate-Based Explosives: Developing highly sensitive detection methods for explosives using electron-rich Ni porphyrins, this application serves crucial roles in safety and security fields, demonstrating the versatility of peroxide tests (Brockmann et al., 2024).


Linkage

Replaces: 16974-1; 16974

Analysis Note

function control: conforms
Assay
- standard 1:
1.8 - 2.2 mg/l
- standard 2: 6.3 - 7.7 mg/l
- standard 3: 13.5 - 16.5 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about disinfection control and how to select the right disinfectant among the various options available in its methods and instrumentation based on photometry and reflectometry.

Protocols

Preparation of a standard solution for Hydrogen peroxide

Related Content

Reflectometric peroxide testing in food packaging materials with RQflex® 20 reflectometer and Reflectoquant® Test Strips.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service