Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.12392

Sigma-Aldrich

Vanadium

powder 99+

Synonym(s):

Vanadium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
MDL number:
UNSPSC Code:
12141749
EC Index Number:
231-171-1

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

mp

1710 °C

density

6.11 g/cm3 at 18.7 °C

bulk density

4000 kg/m3

storage temp.

no temp limit

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Analysis Note

Identity (elemental analysis): conform
Purity (metal base): ≥ 99.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service