Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00824

Supelco

Vanadium(V) oxide

EMPLURA®

Synonym(s):

Vanadium(V) oxide, Vanadic anhydride, Vanadium pentoxide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$1,280.00
1 KG
$3,190.00

$1,280.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$1,280.00
1 KG
$3,190.00

About This Item

Empirical Formula (Hill Notation):
O5V2
CAS Number:
Molecular Weight:
181.88
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-239-8
NACRES:
NA.21

$1,280.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

EMPLURA®

Assay

≥99.0%

form

solid

potency

400-500 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

impurities

≤0.002% Chloride (Cl)
≤0.01% Fe (Iron)
≤0.01% Phosphate (PO4)
≤0.01% Sulfate (SO4)
≤0.01% Total nitrogen (N)

pH

4 (20 °C, 50 g/L in H2O, slurry)

bp

1750 °C/1013 hPa (decomposition)

mp

690 °C

density

3.36 g/cm3 at 20 °C

bulk density

400‑600 kg/m3

storage temp.

2-30°C

SMILES string

[V](=O)(=O)(=O)(=O)(=O)[V]

InChI

1S/5O.2V

InChI key

GNTDGMZSJNCJKK-UHFFFAOYSA-N

Related Categories

Application


  • Demethoxycurcumin in cancer research: This compound is significantly researched for its efficacy in cancer treatment, particularly in cell signaling pathways that govern cell proliferation and apoptosis. Studies indicate its potential in targeting NF-kB pathways, commonly associated with cancer cell survival (Ullah et al., 2024).

Analysis Note

Assay: ≥ 99.0 %
Chloride (Cl): ≤ 0.002 %
Phosphate (PO₄): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.01 %
Total nitrogen (N): ≤ 0.01 %
Fe (Iron): ≤ 0.01 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Lact. - Muta. 2 - Repr. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system, Respiratory Tract

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service