Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09644

Millipore

MAS-100 Iso NT® Dust Cover

stainless steel, Protection of MAS-100 Iso sampling heads from dust

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

stainless steel
stainless steel cover

Agency

non-compendial

feature

autoclavable

packaging

pkg of 1 unit

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100® Iso

shipped in

ambient

storage temp.

no temp limit

General description

MAS-100 ISO NT® Dust Cover, Stainless Steel protects the stainless-steel perforated lids of MAS-100 ISO NT® or MAS-100 ISO MH® from damage and dirt. The design allows easy handling even with isolator gloves. The dust cover is made from stainless steel and can be autoclaved and disinfected with all disinfectants compatible to stainless steel in order to avoid false positive results in isolators, RABS or high-grade cleanrooms.

Application

MAS-100 ISO NT® Dust Cover for collection head prevents cross-contamination for the perforated lid and the agar media, when the air sampler is dormant.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service