Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09093

Sigma-Aldrich

AFB-Fluor

Staining kit for fluorescence-microscopic detection of acid fast bacteria

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130

Quality Level

form

liquid

IVD

for in vitro diagnostic use

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

AFB-Fluor -Staining kit for fluorescence-microscopic detection of acid fast bacteria, is a staining using using fluorescence which allows acid-fast bacteria (AFB) to be easily identified even when their numbers are small. As it has become an established procedure alongside Ziehl-Neelson staining, we now provide a ready-to-use AFB-fluor staining kit. The kit contains three solutions, of which the first solution contains the two fluorescent dyes, Rhodamine B – staining red-orange - and Auramine O – staining yellow-green – which can be used depending on the microscopic filters. Further, the kit supplies a decolorization solution and a potassium permanganate solution that turns the background uniformly black, suppressing troublesome intrinsic fluorescence of the specimen. If required, double staining with AFB-Color modified (Product number 1.00497)(ZN) is possible. AFB-fluor, modified is designed to be used for smears, thougt it can also be used for paraffin sections. The phenol-free kit AFB-fluor phenol-free (Product number 1.01597) can also be used as a user-friendly alternative.
The kit is sufficient for up to 300 applications. It is an IVD product and CE-certified, thus it can be used in diagnostic applications. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Suitability for microscopy: passes test
Mycobacteria: reddish/greenish
Background: dark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service