Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8510-OP

Millipore

Sucrose

OmniPur®

Synonym(s):

Sucrose, α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$56.70
5 KG
$274.00

$56.70


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$56.70
5 KG
$274.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
90825
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.28

$56.70


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

Quality Level

form

crystals (Fine)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

185-187 °C (lit.)

shipped in

ambient

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description



Molecular biology grade.

Specifications:
DNase: None detected
Heavy metals (as Pb): 0.0005% max
Insoluble matter: 0.005% max
Invert sugar: 0.01% max
Iron (Fe): 0.0005% max
Loss on drying: 0.03% max
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Effects of high-temperature stress on gene expression related to photosynthesis in two jujube (Ziziphus jujuba Mill.) varieties.: Investigates the impact of high-temperature stress on photosynthetic gene expression in jujube, revealing adaptive mechanisms that could be linked to sucrose biosynthesis under stress conditions (Yang et al., 2024).

  • Formulation optimization of lyophilized aptamer-gold nanoparticles: Maintained colloidal stability and cellular uptake.: Discusses the optimization of lyophilized aptamer-gold nanoparticle formulations, which could be used for targeted drug delivery systems involving sucrose as a stabilizing agent (Saidi et al., 2024).

  • Preventing too much of a good thing: epigenetic regulation limits sucrose-induced anthocyanin production in Arabidopsis.: Explores how epigenetic mechanisms regulate the anthocyanin biosynthesis pathway in response to sucrose, offering insights into plant stress responses and metabolic regulation (LaFountain, 2024).


Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service