Skip to Content
MilliporeSigma

1.02578

SAFC

Cellvento® Feed-220

Chemically defined cell culture feed

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

storage temp.

2-8°C

General description

Cellvento® Feed-220 is a chemically defined feed formulation containing only components of non-animal origin. The product is intended for use in the development and manufacturing of biotherapeutics in Chinese Hamster Ovary (CHO) cell-based expression systems. Cellvento® Feed-220 contains concentrated amino acids, vitamins, salts, and trace elements and is supplied in dry powder format.

Application

Cellvento® Feed-220 has been designed to support optimal cell growth and performance of CHO-K1 derived suspension cell types, but may be suitable for use with other CHO cell lines. It has been developed to be used as a companion feed supplement with the production medium Cellvento® CHO-220 in fed-batch manufacturing processes. This product is intended for research or further manufacturing but not for human or therapeutic use.

Quantity

Formulated to contain 113 grams of powder per liter of medium.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service