Skip to Content
MilliporeSigma

1.02577

SAFC

Cellvento® CHO-220

Chemically defined cell culture medium

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing

Select a Size

10 L
$1,088.00
100 L
$9,132.00

$1,088.00


Please contact Customer Service for Availability


Select a Size

Change View
10 L
$1,088.00
100 L
$9,132.00

About This Item

UNSPSC Code:
41106213

$1,088.00


Please contact Customer Service for Availability

Quality Level

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

storage temp.

2-8°C

General description

Cellvento® CHO-220 chemically defined cell culture medium has been specially developed for the growth of Chinese Hamster Ovary (CHO) cells and the expression of monoclonal antibodies and recombinant proteins in suspension culture. The formulation is of non-animal origin, chemically defined and contains no hydrolysates or components of unknown composition. Cellvento® CHO-220 medium has been formulated without L-glutamine. It contains hypoxanthine and thymidine, and is available in dry powder form or as ready-to-use medium to fit to different experimental set-ups.

Application

Cellvento® CHO-220 medium and its companion feeds have been designed for use with recombinant CHO-K1 suspension cells, but may also be suitable for other CHO cell lines.
  • Cellvento® CHO-220 medium should be used as an amplification and production medium in fed-batch applications (together with its companion feed product Cellvento® Feed-220).
  • Cellvento® products allow for flexibility in feed and feed supplement optimization of fed-batch processes.
This product is intended for research or further manufacturing but not for human or therapeutic use.

Quantity

Formulated to contain 20.3 grams of powder per liter of medium.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service