Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.00475

Supelco

Phosphate Cell Test

photometric, 0.5-25.0 mg/L (PO4-P), 1.5-76.7 mg/L (PO43-), 1.1-57.3 mg/L (P2O5), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116

product line

Spectroquant®

Quality Level

specific analyte(s)

phosphate

measuring range

0.5-25.0 mg/L (PO4-P)
1.1-57.3 mg/L (P2O5)
1.5-76.7 mg/L (PO43-)

technique(s)

photometry: suitable

detection method

photometric

storage temp.

15-25°C

General description

(o-phosphate) Method: photometric, PMB 0.5 - 25.0 mg/l PO₄-P 1.5 - 76.7 mg/l PO₄³⁻ 1.1 - 57.3 mg/l P₂O₅ Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service