Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S-095

Supelco

PB-22 4-Hydroxypentyl metabolite-D5 (indole-D5) solution

100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H17D5N2O3
Molecular Weight:
379.46
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(OC1=C(N=CC=C2)C2=CC=C1)C3=C([2H])N(CCCC(O)C)C4=C([2H])C([2H])=C([2H])C([2H])=C43

InChI

1S/C23H22N2O3/c1-16(26)7-6-14-25-15-19(18-10-2-3-11-20(18)25)23(27)28-21-12-4-8-17-9-5-13-24-22(17)21/h2-5,8-13,15-16,26H,6-7,14H2,1H3/i2D,3D,10D,11D,15D

InChI key

USLCLHHNJHEBGL-DVHJCUEESA-N

General description

A stable-labeled internal standard of PB-22 4-Hydroxypentyl metabolite, also known as the Omega minus one-OH metabolite of PB-22. The 4-Hydroxypentyl analog is the major urinary metabolite of PB-22, a synthetic analog of JWH-018. The use of PB-22 has been associated with negative side effects such as nausea, vomiting, and seizures.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service