S-102
PB-22 3-carboxyindole metabolite solution
100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
Select a Size
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in acetonitrile
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
[n]1(c2c(c(c1)C(=O)O)cccc2)CCCCC
InChI
1S/C14H17NO2/c1-2-3-6-9-15-10-12(14(16)17)11-7-4-5-8-13(11)15/h4-5,7-8,10H,2-3,6,9H2,1H3,(H,16,17)
InChI key
HAPJUNILBCTRIJ-UHFFFAOYSA-N
General description
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
35.6 °F - closed cup
Flash Point(C)
2.0 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service