Skip to Content
MilliporeSigma
All Photos(1)

Documents

G-020

Supelco

Ganciclovir

1.0 mg/mL (1% 1M HCl in Methanol), certified reference material, Cerilliant®

Synonym(s):

2-Amino-1,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6H-purin-6-one, 9-(1,3-Dihydroxy-2-propoxymethyl)guanine, DHPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N5O4
CAS Number:
Molecular Weight:
255.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL (1% 1M HCl in Methanol)

format

single component solution

storage temp.

2-8°C

SMILES string

NC1=Nc2c(ncn2COC(CO)CO)C(=O)N1

InChI

1S/C9H13N5O4/c10-9-12-7-6(8(17)13-9)11-3-14(7)4-18-5(1-15)2-16/h3,5,15-16H,1-2,4H2,(H3,10,12,13,17)

InChI key

IRSCQMHQWWYFCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ganciclovir is a guanine-analogue, antiviral drug indicated for the treatment of cytomegalovirus (CMV) infection and human herpesvirus 6 (HHV-6). Therapeutic drug monitoring (TDM) of ganciclovir concentrations in patient serum is performed to avoid concentration-related adverse effects such as hepatic and renal toxicity and to evaluate drug adsorption. This Certified Spiking Solution® is suitable for use as starting materials in the preparation of calibrators, controls, or linearity standards for a variety of LC-MS/MS or GC/MS testing methods in TDM, pharmaceutical research and clinical and diagnostic testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Muta. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service