Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G-005

Supelco

Glutethimide solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO2
CAS Number:
Molecular Weight:
217.26
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCC1(CCC(=O)NC1=O)c2ccccc2

InChI

1S/C13H15NO2/c1-2-13(10-6-4-3-5-7-10)9-8-11(15)14-12(13)16/h3-7H,2,8-9H2,1H3,(H,14,15,16)

InChI key

JMBQKKAJIKAWKF-UHFFFAOYSA-N

General description

Glutethimide is a hypnotic sedative sold under the trade name Doriden. Glutethimide has a history of abuse in combination with codeine. This Certified Snap-N-Spike® Solution is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from urine drug testing to forensic analysis and clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Y Aboul-Enein et al.
Biomedical chromatography : BMC, 11(5), 303-306 (1997-10-06)
A series of piperidine-2, 6-dione drugs were enantiomerically resolved on a covalently bonded cellulose 3,5-dimethylphenyl carbamate/10-undecenoate chiral stationary phase (CSP), under normal- or reversed-phase conditions. A single column that can be applied in both modes may be advantageous when considering
Sridevi Kolluri et al.
The Biochemical journal, 392(Pt 1), 173-180 (2005-07-22)
Haem is essential for the health and function of nearly all cells. 5-Aminolaevulinic acid synthase-1 (ALAS-1) catalyses the first and rate-controlling step of haem biosynthesis. ALAS-1 is repressed by haem and is induced strongly by lipophilic drugs that also induce
H Y Aboul-Enein et al.
Die Pharmazie, 59(11), 833-835 (2004-12-14)
The enantiomeric resolution of p-acetylaminoglutethimide and p-nitroglutethimide was achieved on a Ceramospher RU-2 column using methanol as the mobile phase. The flow rates of the mobile phase were 1.0 and 0.5 mL/min for p-acetylaminoglutethimide and p-nitroglutethimide, respectively, with UV detection
Marie-Josée Rocheleau
Electrophoresis, 26(12), 2320-2329 (2005-05-25)
Generic capillary electrophoresis (CE) conditions have been implemented for chiral separations in early pharmaceutical development. The chiral CE separations of several pharmaceutical samples at different stages of development, i.e., discovery, process chemistry, and investigative new drug application, have been obtained
Wojciech Mańkowski et al.
Przeglad lekarski, 59(4-5), 390-391 (2002-08-20)
Suicidal attempt with old (currently unused) drug is described. The suicidal attempts are usually performed with the use of contemporary pharmacotherapeutics. In the report a case of suicidal attempt with old drug Tardyl is presented. Tardyl (Glutethimid) was prescribed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service