Skip to Content
MilliporeSigma
All Photos(1)

Documents

F-023

Supelco

(±)-2-Fluoroamphetamine-D5 (side chain) hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7D5FN · HCl
CAS Number:
Molecular Weight:
194.69
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol (as free base)

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

NC([2H])(C([2H])([2H])[2H])C([2H])C1=C(F)C=CC=C1.[H]Cl

InChI

1S/C9H12FN.ClH/c1-7(11)6-8-4-2-3-5-9(8)10;/h2-5,7H,6,11H2,1H3;1H/i1D3,6D,7D;

InChI key

RAKLYDJAHZCJMS-NINFPRRKSA-N

General description

2-Fluoroamphetamine, or 2-FA, is a stimulant and fluoro analog of amphetamine. Abuse of this club/designer drug has been reported throughout the US and Europe. This internal standard is suitable for numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug monitoring and isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service