Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F-021

Supelco

(±)-2-Fluoromethamphetamine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14FN · HCl
Molecular Weight:
203.68
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

FC1=C(CC(NC)C)C=CC=C1.[H]Cl

InChI

1S/C10H14FN.ClH/c1-8(12-2)7-9-5-3-4-6-10(9)11;/h3-6,8,12H,7H2,1-2H3;1H

InChI key

GTSVNAIEHCXQPM-UHFFFAOYSA-N

General description

A certified solution standard suitable for use as starting material in calibrators and controls for GC/MS or LC/MS methamphetamine testing methods in clinical toxicology, forensic analysis or urine drug testing. A fluoro analog of methamphetamine, 2-Fluoromethamphetamine is sold as a club/designer drug in the illicit market. The drug, also known as 2-FMA, produces stimulant and possibly empathogenic or psychoactive effects.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service