Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERE-032S

Supelco

Ethyl centralite solution

500 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O
CAS Number:
Molecular Weight:
268.35
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1.2 mL

concentration

500 μg/mL in acetonitrile

format

single component solution

storage temp.

2-8°C

Legal Information

Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Zayed et al.
Journal of hazardous materials, 179(1-3), 453-461 (2010-04-07)
Propellants containing nitrate esters need stabilizers to avoid early decomposition or even explosion during storage. Newly prepared malonanilides M1-M5 were tested in stabilizing double-base propellants (DBPs). Their stabilization was compared with the effect of classical stabilizer N,N'-diethyldiphenyl urea (C1) using
María López-López et al.
Talanta, 81(4-5), 1742-1749 (2010-05-06)
In this work, a new approach for the isolation of nitrocellulose from smokeless gunpowders has been developed. A multistep solvent extraction method was needed to purify nitrocellulose contained in gunpowders. For single-base or double-base gunpowders six consecutive solvent extractions were
M A Zayed et al.
Journal of hazardous materials, 73(3), 237-244 (2000-04-07)
Five malonanilide derivatives (M1-M5) were prepared by the reaction of ethyl malonate with aniline derivatives. These compounds were investigated as new stabilizers for double-base propellants (DBPs). The evaluation process has been performed through thermal stability tests, thermal analyses measurements (TGA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service