MilliporeSigma
  • New protocol for the isolation of nitrocellulose from gunpowders: utility in their identification.
New protocol for the isolation of nitrocellulose from gunpowders: utility in their identification.
Talanta (2010-05-06)
María López-López, María Angeles Fernández de la Ossa, Jorge Sáiz Galindo, Jose Luis Ferrando, Alfonso Vega, Mercedes Torre, Carmen García-Ruiz
ABSTRACT
In this work, a new approach for the isolation of nitrocellulose from smokeless gunpowders has been developed. A multistep solvent extraction method was needed to purify nitrocellulose contained in gunpowders. For single-base or double-base gunpowders six consecutive solvent extractions were selected: three extractions with methanol (to remove nitroglycerin, 2,4-dinitrotoluene, ethyl-centralite, diphenylamine, and diphenylamine derivatives); one extraction with dichloromethane (to remove colorants and plasticizers of organic nature); one extraction with methanol (to facilitate a final polar extraction); and one extraction with water (to remove ionic components) were necessary at 35 degrees C. For the triple-base gunpowder studied, eight solvent extractions were needed due to a high concentration of the water-soluble nitroguanidine was present. In addition to the same five initial phases used for the single-base and double-base gunpowders, three water extraction phases at a higher temperature (75 degrees C instead of 35 degrees C) were also needed. A final step to solubilize nitrocellulose in methyl ethyl ketone was used to remove inert components (mainly graphite). Nitrocellulose isolated from these propellants was characterized by Fourier-Transformed Infrared Spectroscopy (FTIR spectroscopy). The same FTIR spectra were observed for nitrocelluloses isolated from different types of gunpowders. A comparison of FTIR spectra of nitrocellulose samples of different nitration degree evidenced that the bands regions most affected by this factor were: 3600-3400cm(-1), corresponding to the stretching vibrations of residual hydroxyl groups; 1200-1000cm(-1), attributed to the valence vibrations nuCO of the glucopyranose cycle; and 750-690cm(-1), assigned to vibrations of the nitrate group. In both cases, the bands appearing in these regions were more pronounced in the spectra of nitrocellulose samples of low nitration degree.
Sigma-Aldrich
1,3-Diethyl-1,3-diphenylurea, 99%