Skip to Content
MilliporeSigma
All Photos(1)

Documents

C-271

Supelco

Chloroquine diphosphate solution

1 mg/mL in methanol: water (1:1) (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Chloroquine diphosphate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26ClN3 · 2H3PO4
CAS Number:
Molecular Weight:
515.86
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

description

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol: water (1:1) (as free base)

format

single component solution

storage temp.

-10 to -25°C

SMILES string

O=P(OO)=O.ClC1=CC=C2C(N=CC=C2NC(CCCN(CC)CC)C)=C1.O=P(OO)=O.[HH].[HH]

InChI

1S/C18H26ClN3.2H3O4P/c1-4-22(5-2)12-6-7-14(3)21-17-10-11-20-18-13-15(19)8-9-16(17)18;2*1-5(2,3)4/h8-11,13-14H,4-7,12H2,1-3H3,(H,20,21);2*(H3,1,2,3,4)

InChI key

QKICWELGRMTQCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Chloroquine, sold under trade names such as Aralen, Avloclor among others, is an aminoquinoline that has antimalarial and anticancer activities. Chloroquine is being studied to treat COVID-19 as of 2020. A Snap-N-Spike® reference solution applicable for use in pharma research and clinical toxicology analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service