Skip to Content
MilliporeSigma
All Photos(1)

Documents

860740L

Avanti

18:1 SM (d18:1/18:1)-d9

Avanti Research - A Croda Brand 860740L-1MG860746L-1MG

Synonym(s):

N-oleoyl-D-erythro-sphingosylphosphorylcholine-d9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H72D9N2O6P
CAS Number:
Molecular Weight:
738.12
UNSPSC Code:
12352100
NACRES:
NA.12

Assay

>99% (TLC)

form

solution

packaging

package of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand 860740L-1MG
860746L-1MG

concentration

1 mg/mL

technique(s)

mass spectrometry (MS): suitable

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Glass Sealed Ampule (860740L-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tong Liu et al.
Expert review of proteomics, 17(3), 233-242 (2020-04-21)
Background: Colorectal cancer (CRC) represents a third leading cause of cancer-related death worldwide. The reliable diagnostic biomarkers for detecting CRC at early stage is critical for decreasing the mortality.Method: A conjunctive lipidomic approach was employed to investigate the differences in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service