Skip to Content
MilliporeSigma
All Photos(2)

Documents

855777P

Avanti

20:0 Lyso PC

1-arachidoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-eicosanoyl-sn-glycero-3-phosphocholine; PC(20:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H58NO7P
CAS Number:
Molecular Weight:
551.74
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 200 mg (855777P-200mg)
pkg of 1 × 25 mg (855777P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855777P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCCCCCCCC)=O

Application

20:0 Lyso PC has been used as an internal standard for the quantification of serum cholesterol and triacylglycerol using liquid chromatography-mass spectrometry (LC-MS) preceded by single-step liquid-liquid extraction method.

Packaging

5 mL Amber Glass Screw Cap Vial (855777P-200mg)
5 mL Amber Glass Screw Cap Vial (855777P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum lipidome analysis of healthy beagle dogs receiving different diets
Boretti FS, et al.
Metabolomics, 16(1), 1-1 (2020)
Kelsey B Law et al.
Autophagy, 13(5), 868-884 (2017-05-20)
Peroxisome biogenesis disorders (PBDs) are metabolic disorders caused by the loss of peroxisomes. The majority of PBDs result from mutation in one of 3 genes that encode for the peroxisomal AAA ATPase complex (AAA-complex) required for cycling PEX5 for peroxisomal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service