Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43951

Sigma-Aldrich

Adipyl chloride

for synthesis

Synonym(s):

Adipyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$99.90
250 ML
$380.00

$99.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$99.90
250 ML
$380.00

About This Item

Empirical Formula (Hill Notation):
C6H8Cl2O2
CAS Number:
Molecular Weight:
183.03
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-876-4
NACRES:
NA.22

$99.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

transition temp

flash point 112 °C (External MSDS)

density

1.259 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

ClC(=O)CCCCC(=O)Cl

InChI

1S/C6H8Cl2O2/c7-5(9)3-1-2-4-6(8)10/h1-4H2

InChI key

PWAXUOGZOSVGBO-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 95.0 %
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service