Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850446P

Avanti

14:0-18:0 PC

1-myristoyl-2-stearoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-tetradecanoyl-2-octadecanoyl-sn-glycero-3-phosphocholine; MSPC; PC(14:0/18:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$190.55
200 MG
$518.09

$190.55


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$190.55
200 MG
$518.09

About This Item

Empirical Formula (Hill Notation):
C40H80NO8P
CAS Number:
Molecular Weight:
734.04
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$190.55


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (850446P-200mg)
pkg of 1 × 25 mg (850446P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850446P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O)=O

InChI

1S/C40H80NO8P/c1-6-8-10-12-14-16-18-19-20-21-23-25-27-29-31-33-40(43)49-38(37-48-50(44,45)47-35-34-41(3,4)5)36-46-39(42)32-30-28-26-24-22-17-15-13-11-9-7-2/h38H,6-37H2,1-5H3/t38-/m1/s1

InChI key

TYAQXZHDAGZOEO-KXQOOQHDSA-N

General description

Phosphatidylcholine (PC) is one of the most important constituent of colon′s mucosal layer.[1] 14:0-18:0 PC (1-myristoyl-2-stearoyl-sn-glycero-3-phosphocholine) is an asymmetrical lipid with long alkyl side chain.[2]

Application

14:0-18:0 PC (1-myristoyl-2-stearoyl-sn-glycero-3-phosphocholine) may be used:
  • in lipid bilayers to study its miscibility with d-erythro-N-palmitoyl-sphingomyelin (16:0-SM) using differential scanning calorimetry (DSC)[3]
  • as an asymmetrical lipid component in liposomes to study the impact of bilayer area and lipid geometry and as liposomal solubilizing agents[2]
  • to prepare a low temperature-sensitive liposome for thrombolytic studies[4]

Biochem/physiol Actions

Phosphatidylcholine (PC) forms a hydrophobic surface in the mucus by acting as a surfactant resulting in the inhibition of penetrance of bacteria.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (850446P-200mg)
5 mL Amber Glass Screw Cap Vial (850446P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory Bowel Disease
Integrative Medicine, 501-516 (2018)
Vishal Saxena et al.
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group, 31(1), 67-73 (2015-03-15)
Clinical efficacy of thrombolytic drugs is limited by lack of specific delivery and requires large therapeutic doses which increase toxicity. Encapsulating these drugs in temperature-sensitive liposomes and applying hyperthermia to deliver thrombolytic agents locally to thrombus might theoretically favourably alter
M Habib Ali et al.
International journal of pharmaceutics, 453(1), 225-232 (2012-07-07)
Liposomes are well recognised for their ability to improve the delivery of a range of drugs. More commonly they are applied for the delivery of water-soluble drugs, but given their structural attributes, they can also be employed as solubilising agents
Van Du Nguyen et al.
ACS applied materials & interfaces, 12(9), 10130-10141 (2020-02-12)
Although great efforts have been undertaken to develop a nanoparticle-based drug delivery system (DDS) for the treatment of solid tumors, the therapeutic outcomes are still limited. Immune cells, which possess an intrinsic ability to phagocytose nanoparticles and are recruited by
Bohdana Térová et al.
Biochimica et biophysica acta, 1667(2), 182-189 (2004-12-08)
In this study we have used differential scanning calorimetry (DSC) to study the miscibility of different saturated phosphatidylcholines (PCs) with D-erythro-N-palmitoyl-sphingomyelin (16:0-SM). Information about the miscibility was obtained by observing the thermotropic phase behavior of binary mixtures of saturated PCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service