MilliporeSigma
  • Miscibility of acyl-chain defined phosphatidylcholines with N-palmitoyl sphingomyelin in bilayer membranes.
Miscibility of acyl-chain defined phosphatidylcholines with N-palmitoyl sphingomyelin in bilayer membranes.
Biochimica et biophysica acta (2004-12-08)
Bohdana Térová, J Peter Slotte, Thomas K M Nyholm
ABSTRACT
In this study we have used differential scanning calorimetry (DSC) to study the miscibility of different saturated phosphatidylcholines (PCs) with D-erythro-N-palmitoyl-sphingomyelin (16:0-SM). Information about the miscibility was obtained by observing the thermotropic phase behavior of binary mixtures of saturated PCs and 16:0-SM. The results obtained showed that PC miscibility in 16:0-SM was markedly affected by the PC acyl-chain composition. According to phase diagrams prepared from DSC data and the mid-transition temperatures of the main phase transition, the PC which formed the most ideal mixture with 16:0-SM was di-14:0-PC. However, the cooperativity of the main transition in PC/16:0-SM bilayers increased until the average acyl-chain length in the PC reached 15 carbons. Based on the criteria of the most ideal miscibility or the highest cooperativity of the main transition, we conclude that di-14:0-PC, 15:0/15:0-PC, and 14:0/16:0-PC interacted most favorably with 16:0-SM in bilayer membranes. Di-16:0-PC, to which 16:0-SM is often compared in biophysical studies, showed much less ideal miscibility.
Product Description
Avanti
14:0-18:0 PC, 1-myristoyl-2-stearoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
14:0-16:0 PC, 1-myristoyl-2-palmitoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
14:0-16:0 PC, 1-myristoyl-2-palmitoyl-sn-glycero-3-phosphocholine, powder
Avanti
14:0-18:0 PC, 1-myristoyl-2-stearoyl-sn-glycero-3-phosphocholine, powder