Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

840537L

Avanti

18:1 SQDG(d18)

Avanti Research - A Croda Brand

Synonym(s):

Sulfoquinovosyldioleoylglycerol (d18)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$743.00

$743.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$743.00

About This Item

Empirical Formula (Hill Notation):
C45H67D18NO12S
CAS Number:
Molecular Weight:
882.33
UNSPSC Code:
12352211
NACRES:
NA.21

$743.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

(DCM:Methanol (1:1) solution)

packaging

pkg of 1 × 1 mg 840537L-1MG

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

shipped in

dry ice

storage temp.

−20°C

Application

lipidomics

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service