Skip to Content
MilliporeSigma
All Photos(1)

Documents

840534L

Avanti

18:1 DGDG(d18)

Avanti Research - A Croda Brand

Synonym(s):

1,2-Dioleoyl-3-β-D-galactopyranosyl-sn-glycerol (d18), Digalactosyldioleoylglycerol (d18)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H74D18O15
Molecular Weight:
963.38
UNSPSC Code:
12352211
NACRES:
NA.21

Assay

>99% (TLC)

form

(DCM:Methanol (1:1) solution)

packaging

pkg of 1 × 1 mg (840534L-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

shipped in

dry ice

storage temp.

−20°C

Application

Lipidomics

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service