Skip to Content
MilliporeSigma
All Photos(1)

Documents

792455C

Avanti

16:0-18:1(n7) PC - IsoPure

Avanti Research - A Croda Brand

Synonym(s):

1-palmitoyl-2-(11Z-octadecenoyl)-glycero-3-phosphocholine (IsoPure)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8P
CAS Number:
Molecular Weight:
760.08
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mL

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL in chloroform

technique(s)

mass spectrometry (MS): suitable

application(s)

lipidomics
metabolomics

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCC/C=C\CCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

Packaging

5 mL Clear Glass Sealed Ampule (792455C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service