792454C
Avanti
16:0-18:1(n10) PC - IsoPure
Avanti Research™ - A Croda Brand
Synonym(s):
1-palmitoyl-2-(8Z-octadecenoyl)-sn-glycero-3-phosphocholine (IsoPure)
Select a Size
Select a Size
About This Item
Recommended Products
Assay
>99% (TLC)
form
chloroform solution
packaging
pkg of 1 × 1 mL
manufacturer/tradename
Avanti Research™ - A Croda Brand
concentration
10 mg/mL in chloroform
technique(s)
mass spectrometry (MS): suitable
application(s)
lipidomics
metabolomics
lipid type
phospholipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCC/C=C\CCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O
InChI key
FNCGJGDCVBZNMN-GPIBEQEUSA-N
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
Target Organs
Central nervous system, Liver,Kidney
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service