Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z743857

ART® Non-Filtered Tip

HLT 250, size 200 μL, bulk pack, non-sterile

Synonym(s):

aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

sterility

non-sterile

feature

HLT 250
bulk pack

packaging

pkg of 10X1000 ea

manufacturer/tradename

Molecular BioProducts 3550

technique(s)

PCR: suitable

size

200 μL

Looking for similar products? Visit Product Comparison Guide

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service