Z743738
ART® Low Retention Barrier Tip
ART 200, size 200 μL, racked, sterile
Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
sterile
feature
ART 200
racked
packaging
pkg of 5X960 ea
manufacturer/tradename
Molecular BioProducts 2069-05
technique(s)
PCR: suitable
size
200 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- ART® low retention barrier tips are designed to maximize sample recovery.
- Tip′s hydrophobic inner surface prevents sample loss, facilitates sample accuracy and reproducibility.
- Clear tips provide sample clarity.
- Self-sealing technology prevents cross-contamination.
- Certified RNase, DNase, ATP, endotoxin, and pyrogen free.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service