CLS1700100
Corning® coliform water test sample container, sterile
with sodium thiosulfate tablet, 120 mL polypropylene bottle, graduated, sterile, 100/cs
Synonym(s):
sterile sample bottles
Select a Size
$161.00
Available to ship onApril 24, 2025Details
Select a Size
About This Item
$161.00
Available to ship onApril 24, 2025Details
Recommended Products
packaging
case of 100
pack of 100
manufacturer/tradename
Corning 1700-100
Related Categories
General description
- Well defined EPA Fill Line for 100 mL
- Reinforced custody string
- Large thumb tabs to reduce contamination and ease for opening
- 2 thumb taps on either side of the locking mechanism
- 1 in. headspace between water level and underside of closed lid
Features and Benefits
- Corning® coliform water test sample container, sterile, is used in the drinking water testing for the presence of coliform bacteria and other microbiological contaminants.
- Pure polypropylene with Capacity: 120 mL total capacity.
- Provided with a lid to reduce the chance of contamination.
- Locking arrow assures sterility has not been compromised.
- Inclusive of a sodium thiosulfate tablet in container.
- Leak proof.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)