Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743689

ART® barrier tip empty hinged rack, ART essentials kit

Synonym(s):

barrier tip storage, tip rack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

packaging

pkg of 4X1152 ea

manufacturer/tradename

Molecular BioProducts 2921-HR

technique(s)

PCR: suitable

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service