Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z699489

Dilution bottle

Phosphate Buffer, Magnesium Chloride, volume 99 mL

Synonym(s):

diltuion bottles, dilution bottle, phosphate buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

packaging

pkg of 72 ea

manufacturer/tradename

Maine Mfg 10498505

capacity

99 mL

volume

99 mL

Looking for similar products? Visit Product Comparison Guide

General description

Phosphate Buffer with Magnesium Chloride is used as the diluent for the preparation of dilutions in plate counts in the dairy and food industries. It is recommended by the APHA for the recovery of injured microorganisms from dairy and food samples. Contains deionized water, monopotassium phosphate, and magnesium chloride.
Whatman sterile dilution bottles are designed for sample dilution of water, dairy products, foods, and pharmaceuticals prior to microbiological testing. Final pH for all solutions is 7.2 pH ± 0.2 pH at 25°C. The buffers are supplied in an easy open, flip-top, plastic container with a tamper-evident seal.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service