Skip to Content
MilliporeSigma
All Photos(2)

Documents

Z676799

ART® barrier tip

maximum volume 1000 μL, Tip design ART 1000XL, Hinged rack, low retention: no

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier
low retention: no
Hinged rack
Tip design ART 1000XL

packaging

case of 3072 ea (96 tips/tray, 8 trays/pack, 4 packs/case)

manufacturer/tradename

Molecular BioProducts 2179-HR

technique(s)

PCR: suitable

maximum volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

The self-sealing ART barrier tips are supplied in environmentally friendly packaging. The sterile, barrier tip reload system combines individually packaged blister tubs made of recycled plastic and new hinged racks. The push down blister tub easily inserts the ART barrier tips into the hinged racks providing stackable, space saving racks that offer easy, one-handed use. Empty hinged rack part numbers allow for easy conversion to the ART Reload System.

Manufactured according to the industry′s highest quality standards, ART tips are certified as RNase, DNase and Pyrogen free, and are ideal for the pipetting of radioactive samples, nucleic amplification procedures, or any applications where critical sample handling is required.

Available as reload inserts consisting of sterile pre-loaded blister tubs of 96 tips with or without hinged racks. The empty hinged racks are also available separately.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service