Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z569755

ART® self-sealing barrier pipette tips

ART 1000E, volume range 100-1000 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01

material

colorless
polypropylene

sterility

sterile

feature

DNase free
RNase free
barrier
endotoxin free

packaging

pack of 800 ea

manufacturer/tradename

Molecular BioProducts 2079E

technique(s)

PCR: suitable

maximum volume

1 mL

volume range

100-1000 μL

Looking for similar products? Visit Product Comparison Guide

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service