Documents
Z653330
Clips for Quickfit® flat-flange reaction flask lids
For 500mL
Synonym(s):
JC75F
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
stainless steel
label
D
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Quickfit JC75F
availability
available only in EU
flange bore
75 mm
General description
Designed for securing flat flange joints, these clips have a hinged ′spring wire′ design that tightens using a wing nut.
Legal Information
Quickfit is a registered trademark of SciLabware Limited