Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z639060

Bollé Viper safety eyeshields

clear polycarbonate lens

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
46181802
NACRES:
NB.35

material

clear polycarbonate lens

grade

CE Compliant

manufacturer/tradename

Bollé VIPCI

availability

not available in USA and Canada

color

frame, electric blue

Looking for similar products? Visit Product Comparison Guide

General description

  • Anti-scratch
  • Adjustable temples

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service