Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z205400

Sigma-Aldrich

Safety glasses dispenser

W × H × D 8 in. × 20 in. × 4 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$523.00

$523.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$523.00

About This Item

UNSPSC Code:
46181802
NACRES:
NB.35

$523.00


Please contact Customer Service for Availability

material

clear acrylic

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F24876-0000

W × H × D

8 in. × 20 in. × 4 in.

General description

Clear acrylic dispenser allows workers to know how many glasses are available at a glance. Glasses held in by flip-up access door.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service