Key Documents
Z564001
Büchi® Rotavapor® RII evaporator with jack and water bath
joint: ST/NS 29/32, plastic-coated glass, condenser, diagonal, AC/DC input 230 - 240 V AC
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
material
plastic-coated glass
Joint
joint: ST/NS29/32
feature
CE compliant
CSA approved
condenser diagonal
manufacturer/tradename
Büchi 2411A1
General description
The Rotavapor RII fullfills exactly the fundamental needs of classical laboratory applications in teaching and research, practically and dependably, without compromising quality or safety. Available in three different glassware assemblies, all supplied with a standard 24/40 joint (US) or 29/32 (Europe) on the evaporator flask and a 35/20 ball joint on the receiving flask, and a choice of either standard glass or plastic-coated glass.
Model R II features:
  • Continuous vapor duct and a PTFE composite vacuum seal for long service life and effective sealing
  • Small footprint and robust design in powder-coated aluminum
  • Manual quick-action jack
  • Heating bath for temperatures of 0 to 95°C, quick-release electrical connection for easy removal
  • Process samples from 50 mL to 4 L
Evaporator with diagonal condenser assembly
For simple distillation of solvents and general use. Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
Evaporator with vertical condenser assembly
For standard distillations and for high-boiling solvents (>100°C). Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
Evaporator with cold-trap condenser assembly
Two-piece cold-trap condenser style is ideal for low-boiling solvents. Uses mechanical refrigeration or dry ice and acetone.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Rotavapor is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)