Key Documents
Z549517
Sigma-Aldrich
Stopcock Adapter
straight PTFE stopcock, joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Pricing and availability is not currently available.
material
straight PTFE stopcock
Joint
joint: ST/NS14/20
General description
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)