Documents
Z544426
Molymod® bonding links
V-bond, gray
About This Item
UNSPSC Code:
60104505
NACRES:
NB.17
packaging
pkg of 25
pkg of 4 pkg
manufacturer/tradename
Molymod
L
3 mm
color
gray
General description
The bond lengths stated in the table are measured from atom to atom. Bonding links are available in various colors and may be used to represent different bond types. For example, gray links can represent covalent bonds, while purple or white links can represent ionic bonds.
Legal Information
Molymod is a registered trademark of Spiring Enterprises Ltd.