Documents
Z406295
Sigma-Aldrich
Sure/Seal bottles
capacity 125 mL, amber plastic-coated glass
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
amber plastic-coated glass
round bottle
Joint
joint threaded neck
feature
without cap
packaging
pkg of 6 ea
manufacturer/tradename
Sigma-Aldrich
capacity
125 mL
General description
Order overcaps, steel crown caps, and PTFE-faced liners separately.
Application
Bottles are used to store chemicals, solvents, and other liquids. Amber-tinted glass is recommended for light-sensitive materials.
Legal Information
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC