Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z112976

Sigma-Aldrich

Crown-cap crimper

H × diam. 21 1/2 in. × 12 in.

Synonym(s):

Bottle cap crimper, Crown cap sealer

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,370.00

$2,370.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$2,370.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$2,370.00


Please contact Customer Service for Availability

H × diam.

21 1/2 in. × 12 in.

Application

Crown-cap crimper may be used to cap the amber-colored glass bottles containing solvent with an Aldrich® Sure-Seal polytetrafluoroethylene (PTFE) disk and a metal crown cap.[1]

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie J Conn et al.
Journal of natural products, 78(1), 120-124 (2014-12-18)
The first total synthesis of the potent and selective human blood coagulation factor XIa inhibitor clavatadine A (1) is described. Direct, early-stage guanidinylation enabled rapid, convergent access to an immediate clavatadine A precursor. Concomitant lactone hydrolysis and guanidine deprotection with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service