Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z317772

Duran® vacuum desiccator replacement lids

knobbed, nominal diameter 250 mm , DN

Synonym(s):

244106608

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.43

manufacturer/tradename

DURAN®
Schott

availability

not available in USA and Canada

nominal diameter

250 mm , DN

Looking for similar products? Visit Product Comparison Guide

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service