Key Documents
Z301124
Quickfit® expansion adapter
top female joint: ST/NS 29/32, bottom male joint: ST/NS 14/23
Synonym(s):
XA41
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
female joint: ST/NS top29/32
male joint: ST/NS bottom14/23
manufacturer/tradename
Quickfit XA 41
availability
not available in USA and Canada
not available in USA and Canada
General description
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)