Key Documents
Z766860
Microflex NeoPro® EC powder-free chloroprene gloves
size L
Synonym(s):
disposable glove
Select a Size
50 EA
$31.30
500 EA
$313.00
$31.30
Please contact Customer Service for Availability
Select a Size
Change View
50 EA
$31.30
500 EA
$313.00
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
$31.30
Please contact Customer Service for Availability
material
(chloroprene gloves)
green
size
L
description
ambidexterous
textured fingers
feature
CE compliant
disposable
latex free
powder-free
packaging
pack of 50 ea (boxes of 50)
manufacturer/tradename
Microflex NEC-288-L
L
11.8 in. , cuff
thickness
6.3 mil , palm
8.3 mil , fingers
General description
Featureing an extended cuff, NeoPro® EC is the longest and thickest non-latex glove produced by Microflex. It uses a polychloroprene formulation to provide a secure grip along with exceptional protection from biological and chemical hazards inwet or dry conditions.
Legal Information
NeoPro is a registered trademark of CIT Healthcare LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)