Key Documents
Z266043
Supelco
Aluminum multi-plate racks
For use with 10 cm × 10 cm plates
About This Item
UNSPSC Code:
41115711
eCl@ss:
32110504
technique(s)
thin layer chromatography (TLC): suitable
General description
For use with Latch-Lid developing chambers.
Legal Information
Latch-Lid is a trademark of General Glassblowing, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)