Key Documents
Z171514
Sigma-Aldrich
Series A gas syringe
capacity 250 μL
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
bevel tip stainless steel needle
needle size
2 in.
feature
needle bevel tip
manufacturer/tradename
VICI 010031
capacity
250 μL
needle L
2.25 in.
General description
Same as series A-2 gas syringes above, but with turn valve instead of push-button valve.
Choose from one of the most recent versions:
Certificates of Analysis (COA)