Key Documents
Z107182
Sigma-Aldrich
Base plates
Unthreaded
Select a Size
1 EA
$38.20
10 EA
$292.00
$38.20
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$38.20
10 EA
$292.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
$38.20
Please contact Customer Service for Availability
Related Categories
General description
Fits ½ in. lattice rods. Flat base.
These base plates are of a non-ferrous alloy, two inches in diameter, 16 mm high, with set screw for fastening lattice frame rods. Provided without mounting screws. Plate has four holes for attaching to bench, floor or wall.
Choose from one of the most recent versions:
Certificates of Analysis (COA)