Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WHA67245002

Whatman® Polydisc AS in-line filter for aqueous solutions

pore size 0.2 μm

Synonym(s):

Whatman filter, Whatman filters, Z744336, syringe filter

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$409.00

$409.00


Available to ship onApril 25, 2025Details



Select a Size

Change View
1 PKG
$409.00

About This Item

UNSPSC Code:
41104922
NACRES:
NB.24

$409.00


Available to ship onApril 25, 2025Details


material

polyethersulfone membrane
polypropylene housing

sterility

non-sterile

packaging

pkg of 10 ea

manufacturer/tradename

Whatman 6724-5002
Whatman Article No. 28420225 (US reference)

diam.

50 mm

pore size

0.2 μm

Looking for similar products? Visit Product Comparison Guide

General description

Whatman Polydisc AS (Aqueous Solution) in-line filters from Cytiva feature high-throughput PES membrane with a glass microfiber prefilter. Available irradiated or nonsterile.

Polyethersulfone membranes for aqueous solutions

AS Polydisc filters use polyethersulfone membranes, which are low protein binding and surfactant free, making them the recommended membrane for many applications such as buffer or reagent filtration.

These membranes have been demonstrated to contribute low levels of extractables which might otherwise interfere with sample analysis. Polydisc AS also incorporates a glass microfiber prefilter to handle heavy particulates and extend membrane life.

Application

Tissue culture media, Reagent preparation, Particle counting solutions, Pharmaceutical preparations

Features and Benefits

  • Whatman filters with polyethersulfone membranes have low protein binding and are surfactant-free
  • GMF prefilter removes heavy particulates, increasing membrane longevity
  • Gamma irradiated, so there are no ethylene oxide (EtO) residuals
  • Barbed hose connections fit multiple tubing sizes
  • Integrity-testable with the bubble point method
  • Lightweight design prevents tubing collapse
  • Recyclable after use.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service